Assay Details
Target Gene Details
Entrez Gene ID: | 399474 |
Gene Name: | transmembrane protein 200B |
Gene Aliases: |
TTMB |
Location: |
Chr.1:29119425-29129138 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM200B | NM_001003682.3 | 2 | 500 | NP_001003682.1 |
| NM_001171868.1 | 2 | 584 | NP_001165339.1 | |
| XM_011541457.2 | 2 | 625 | XP_011539759.1 | |
| AL110282.1 | 1 | 791 | CAH10727.1 | |
| BC064375.1 | 2 | 486 | ||
| BC064538.1 | 1 | 425 | AAH64538.1 | |
| BC082989.1 | 2 | 500 | AAH82989.1 | |
| BC110336.1 | 2 | 584 | AAI10337.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509091 | Chr.1:29103220 - 29163899 on Build GRCh38 | Insertion |
|

Genomic Map