Assay Details
Target Gene Details
Entrez Gene ID: | 83854 |
Gene Name: | angiopoietin like 6 |
Gene Aliases: |
AGF, ARP5 |
Location: |
Chr.19:10092337-10106194 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPTL6 | NM_031917.2 | NP_114123.2 | ||
| XM_005260091.3 | 1 | 141 | XP_005260148.1 | |
| XM_011528347.2 | XP_011526649.1 | |||
| XM_011528348.2 | XP_011526650.1 | |||
| XM_011528349.2 | XP_011526651.1 | |||
| XM_011528350.2 | XP_011526652.1 | |||
| XM_017027347.1 | XP_016882836.1 | |||
| AB054064.1 | BAB91248.1 | |||
| AF230330.1 | AAK06404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3643648 | Chr.19:10101662 - 10103956 on Build GRCh38 | Loss |
|
| nsv578610 | Chr.19:10102342 - 10103112 on Build GRCh38 | Loss |
|
| esv29826 | Chr.19:10101499 - 10103738 on Build GRCh38 | Loss |
|
| esv2675304 | Chr.19:10101623 - 10103908 on Build GRCh38 | Deletion |
|
| nsv833743 | Chr.19:10077649 - 10251913 on Build GRCh38 | Loss |
|
| esv3556016 | Chr.19:10101510 - 10103970 on Build GRCh38 | Deletion |
|
| dgv6266n54 | Chr.19:10102229 - 10103354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map