Assay Details
Target Gene Details
Entrez Gene ID: | 2587 |
Gene Name: | galanin receptor 1 |
Gene Aliases: |
GALNR, GALNR1 |
Location: |
Chr.18:77250052-77270142 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALR1 | NM_001480.3 | 1 | 615 | NP_001471.2 |
| XM_017025691.1 | 1 | 615 | XP_016881180.1 | |
| AY541036.1 | 1 | 118 | AAS47032.1 | |
| BC095530.1 | 1 | 123 | AAH95530.1 | |
| L34339.1 | 1 | 118 | AAA50767.1 | |
| U23854.1 | 1 | 118 | AAB60356.1 | |
| U53511.1 | 1 | 118 | AAC51936.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583165 | Chr.18:77247784 - 77255913 on Build GRCh38 | Loss |
|
| esv2758740 | Chr.18:77206470 - 77465982 on Build GRCh38 | Loss |
|
| esv2717388 | Chr.18:76288671 - 78160297 on Build GRCh38 | Deletion |
|
| esv3643201 | Chr.18:77032848 - 77365699 on Build GRCh38 | Loss |
|
| dgv3407n100 | Chr.18:77173514 - 78799796 on Build GRCh38 | Gain |
|
| nsv2364 | Chr.18:77235963 - 77268176 on Build GRCh38 | Insertion |
|
| dgv1005e212 | Chr.18:77238951 - 77255913 on Build GRCh38 | Loss |
|
| dgv196n21 | Chr.18:77242359 - 77296076 on Build GRCh38 | Gain |
|
| esv2751779 | Chr.18:77171456 - 77646342 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map