Assay Details
Target Gene Details
Entrez Gene ID: | 2309 |
Gene Name: | forkhead box O3 |
Gene Aliases: |
AF6q21, FKHRL1, FKHRL1P2, FOXO2, FOXO3A |
Location: |
Chr.6:108559823-108684769 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXO3 | NM_001455.3 | 1 | 109 | NP_001446.1 |
| NM_201559.2 | NP_963853.1 | |||
| AF032886.1 | 1 | 690 | AAC39592.1 | |
| AK092357.1 | ||||
| BC020227.1 | AAH20227.1 | |||
| BC021224.2 | AAH21224.1 | |||
| BC068552.1 | 1 | 109 | AAH68552.1 | |
| CA389775.1 | 1 | 37 | ||
| CD101760.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv10904n54 | Chr.6:108559308 - 108561930 on Build GRCh38 | Loss |
|
| nsv823817 | Chr.6:108556562 - 108562652 on Build GRCh38 | Gain |
|
| nsv604482 | Chr.6:108559788 - 108561930 on Build GRCh38 | Gain |
|
| nsv604476 | Chr.6:108556849 - 108561930 on Build GRCh38 | Gain |
|
| nsv1017621 | Chr.6:108524796 - 108569702 on Build GRCh38 | Gain |
|
| nsv830767 | Chr.6:108476568 - 108667716 on Build GRCh38 | Gain |
|
| esv22566 | Chr.6:108557767 - 108562009 on Build GRCh38 | Loss |
|
| nsv604477 | Chr.6:108558488 - 108561930 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map