Assay Details
Target Gene Details
Entrez Gene ID: | 26470 |
Gene Name: | seizure related 6 homolog like 2 |
Gene Aliases: |
BSRPA, PSK-1 |
Location: |
Chr.16:29871159-29899573 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEZ6L2 | NM_001114099.2 | NP_001107571.1 | ||
| NM_001114100.2 | NP_001107572.1 | |||
| NM_001243332.1 | NP_001230261.1 | |||
| NM_001243333.1 | NP_001230262.1 | |||
| NM_012410.3 | NP_036542.1 | |||
| NM_201575.3 | NP_963869.2 | |||
| XM_005255252.2 | XP_005255309.1 | |||
| XM_017023135.1 | XP_016878624.1 | |||
| AB105376.1 | ||||
| AF131749.1 | ||||
| AJ245820.1 | CAB57950.1 | |||
| AJ245821.1 | CAB57951.1 | |||
| AJ245822.1 | CAB57952.1 | |||
| AK027407.1 | ||||
| AK027676.1 | ||||
| AK074998.1 | BAC11345.1 | |||
| AK293675.1 | ||||
| AK299103.1 | ||||
| AY358404.1 | AAQ88770.1 | |||
| BC000567.2 | AAH00567.1 | |||
| KJ534938.1 | ||||
| KJ534939.1 | ||||
| KJ534940.1 | ||||
| KJ534941.1 | ||||
| KJ534988.1 | ||||
| KJ534989.1 | ||||
| KJ535068.1 | ||||
| KJ535092.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv571818 | Chr.16:29861040 - 29874126 on Build GRCh38 | Loss |
|

Genomic Map