Assay Details
Target Gene Details
Entrez Gene ID: | 92335 |
Gene Name: | STE20-related kinase adaptor alpha |
Gene Aliases: |
LYK5, NY-BR-96, PMSE, STRAD, Stlk |
Location: |
Chr.17:63702832-63741970 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRADA | NM_001003786.2 | 9 | 1100 | NP_001003786.1 |
| NM_001003787.2 | 11 | 1211 | NP_001003787.1 | |
| NM_001003788.2 | 10 | 1182 | NP_001003788.1 | |
| NM_001165969.1 | 9 | 1124 | NP_001159441.1 | |
| NM_001165970.1 | 9 | 1079 | NP_001159442.1 | |
| NM_153335.5 | 9 | 1100 | NP_699166.2 | |
| XM_005257797.2 | 11 | 1062 | XP_005257854.1 | |
| XM_005257798.2 | 9 | 1024 | XP_005257855.1 | |
| XM_005257799.2 | 10 | 1033 | XP_005257856.1 | |
| XM_005257800.2 | 11 | 1086 | XP_005257857.1 | |
| XM_005257801.4 | 11 | 1412 | XP_005257858.1 | |
| XM_005257803.4 | 10 | 1240 | XP_005257860.1 | |
| XM_011525466.2 | 11 | 1086 | XP_011523768.1 | |
| XM_011525467.2 | 11 | 1113 | XP_011523769.1 | |
| XM_017025312.1 | 9 | 975 | XP_016880801.1 | |
| XM_017025313.1 | 10 | 1057 | XP_016880802.1 | |
| XM_017025314.1 | 9 | 1350 | XP_016880803.1 | |
| AF308302.1 | 11 | 1050 | AAG48269.1 | |
| AK074771.1 | 9 | 966 | BAC11197.1 | |
| AK075005.1 | 9 | 929 | BAC11349.1 | |
| AK128173.1 | 1 | 2021 | ||
| AK293160.1 | 9 | 987 | ||
| AK301331.1 | 9 | 951 | ||
| AL832407.1 | 10 | 995 | CAI46194.1 | |
| AY290821.2 | 9 | 1100 | AAP42280.1 | |
| BC043641.1 | 10 | 1290 | AAH43641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv575854 | Chr.17:63702567 - 63708544 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map