Assay Details
Target Gene Details
Entrez Gene ID: | 199786 |
Gene Name: | family with sequence similarity 129 member C |
Gene Aliases: |
BCNP1 |
Location: |
Chr.19:17523301-17556755 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM129C | NM_001098524.1 | 11 | 1456 | NP_001091994.1 |
| NM_001321826.1 | 10 | 1328 | NP_001308755.1 | |
| NM_001321827.1 | 10 | 1328 | NP_001308756.1 | |
| NM_001321828.1 | 9 | 1166 | NP_001308757.1 | |
| NM_173544.4 | 11 | 1456 | NP_775815.2 | |
| XM_005259813.3 | 10 | 1303 | XP_005259870.1 | |
| XM_005259814.3 | 10 | 1303 | XP_005259871.1 | |
| XM_011527781.2 | 10 | 1303 | XP_011526083.1 | |
| XM_011527786.2 | 10 | 1303 | XP_011526088.1 | |
| XM_011527787.2 | 10 | 1303 | XP_011526089.1 | |
| XM_011527789.1 | 10 | 1303 | XP_011526091.1 | |
| XM_017026453.1 | 10 | 1303 | XP_016881942.1 | |
| XM_017026454.1 | 10 | 1303 | XP_016881943.1 | |
| XM_017026455.1 | 9 | 1175 | XP_016881944.1 | |
| XM_017026456.1 | 9 | 1230 | XP_016881945.1 | |
| XM_017026457.1 | 7 | 880 | XP_016881946.1 | |
| AK074069.1 | 10 | 1278 | BAB84895.1 | |
| AK097121.1 | 10 | 1303 | BAC04955.1 | |
| AK298062.1 | 9 | 1157 | ||
| AK300046.1 | 10 | 1310 | ||
| AK301159.1 | 10 | 1206 | ||
| AY254197.1 | 11 | 1456 | AAO83574.1 | |
| AY254198.1 | 11 | 1456 | AAO83575.1 | |
| AY254199.1 | 11 | 1456 | AAO83576.1 | |
| BC053606.1 | 10 | 1262 | AAH53606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422429 | Chr.19:17529868 - 17579053 on Build GRCh38 | Duplication |
|

Genomic Map