Assay Details
Target Gene Details
Entrez Gene ID: | 2266 |
Gene Name: | fibrinogen gamma chain |
Gene Aliases: |
- |
Location: |
Chr.4:154604134-154612808 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGG | NM_000509.5 | 1 | 102 | NP_000500.2 |
| NM_021870.2 | NP_068656.2 | |||
| AK290824.1 | ||||
| BG567044.1 | ||||
| BX382942.2 | 1 | 142 |
More Information
Set Membership: |
|

Genomic Map