Assay Details
Target Gene Details
Entrez Gene ID: | 9128 |
Gene Name: | pre-mRNA processing factor 4 |
Gene Aliases: |
HPRP4, HPRP4P, PRP4, Prp4p, RP70, SNRNP60 |
Location: |
Chr.9:113275634-113292905 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF4 | NM_001244926.1 | 3 | 401 | NP_001231855.1 |
| NM_001322266.1 | 3 | 401 | NP_001309195.1 | |
| NM_001322267.1 | 3 | 404 | NP_001309196.1 | |
| NM_004697.4 | 3 | 404 | NP_004688.2 | |
| NR_136265.1 | 3 | 404 | ||
| NR_136266.1 | 3 | 401 | ||
| AB209797.1 | 3 | 349 | BAD93034.1 | |
| AF016369.1 | 3 | 354 | AAC51925.1 | |
| AK225056.1 | 3 | 377 | ||
| AK313131.1 | 3 | 346 | ||
| AY572489.1 | 3 | 291 | AAS78572.1 | |
| BC001588.2 | 3 | 344 | AAH01588.1 | |
| BC007424.2 | 3 | 354 | AAH07424.1 | |
| CR457105.1 | 3 | 291 | CAG33386.1 | |
| DA815191.1 | 3 | 401 | ||
| U82756.1 | 3 | 378 | AAC02261.1 |
More Information
Set Membership: |
|

Genomic Map