Assay Details
Target Gene Details
Entrez Gene ID: | 220979 |
Gene Name: | chromosome 10 open reading frame 25 |
Gene Aliases: |
- |
Location: |
Chr.10:44993932-45000910 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf25 | NR_126421.1 |
Target Gene Details
Entrez Gene ID: | 83937 |
Gene Name: | Ras association domain family member 4 |
Gene Aliases: |
AD037 |
Location: |
Chr.10:44959753-44994724 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASSF4 | NM_032023.3 | 11 | 2427 | NP_114412.2 |
| XM_005271833.4 | 11 | 2477 | XP_005271890.1 | |
| XM_005271835.4 | 11 | 2427 | XP_005271892.1 | |
| XM_006718013.3 | 11 | 2522 | XP_006718076.1 | |
| XM_006718014.3 | 10 | 3941 | XP_006718077.1 | |
| XM_011540255.2 | 10 | 2302 | XP_011538557.1 | |
| XM_017016754.1 | 10 | 4067 | XP_016872243.1 | |
| XM_017016755.1 | 10 | 4022 | XP_016872244.1 | |
| XM_017016756.1 | 11 | 2548 | XP_016872245.1 | |
| XM_017016757.1 | 10 | 4093 | XP_016872246.1 | |
| XM_017016758.1 | 9 | 3798 | XP_016872247.1 | |
| AB209446.1 | 10 | 4299 | BAD92683.1 | |
| AL713716.1 | 3 | 1600 | CAD28511.1 | |
| BC032593.1 | 11 | 2409 | AAH32593.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576560 | Chr.10:44916005 - 45054697 on Build GRCh38 | Gain |
|
| nsv526734 | Chr.10:44963037 - 44996606 on Build GRCh38 | Loss |
|
| esv3623079 | Chr.10:44993759 - 45028111 on Build GRCh38 | Loss |
|
| nsv831846 | Chr.10:44826788 - 45060922 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map