Assay Details
Target Gene Details
Entrez Gene ID: | 27165 |
Gene Name: | glutaminase 2 |
Gene Aliases: |
GA, GLS, LGA, hLGA |
Location: |
Chr.12:56470944-56488414 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLS2 | NM_001280796.1 | NP_001267725.1 | ||
| NM_001280797.1 | NP_001267726.1 | |||
| NM_001280798.1 | NP_001267727.1 | |||
| NM_013267.3 | NP_037399.2 | |||
| XM_005268797.1 | XP_005268854.1 | |||
| XM_017019180.1 | XP_016874669.1 | |||
| AF110330.1 | AAF21933.1 | |||
| AF223944.1 | AAF33826.1 | |||
| AK000467.1 | ||||
| AK289471.1 | ||||
| AK294822.1 | ||||
| AK303772.1 | ||||
| BC119767.1 | ||||
| BC119768.1 | ||||
| BI916699.1 | ||||
| DA060302.1 | ||||
| DA180204.1 | ||||
| HM775422.1 | ||||
| HM775423.1 | ||||
| HM775424.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| nsv1052713 | Chr.12:56454252 - 56523818 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map