Assay Details
Target Gene Details
Entrez Gene ID: | 29841 |
Gene Name: | grainyhead like transcription factor 1 |
Gene Aliases: |
LBP32, MGR, NH32, TFCP2L2 |
Location: |
Chr.2:9951663-10002284 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRHL1 | NM_198182.2 | NP_937825.2 | ||
| XM_005246159.3 | XP_005246216.1 | |||
| XM_006711882.2 | XP_006711945.1 | |||
| XM_006711884.3 | XP_006711947.1 | |||
| XM_011510343.2 | XP_011508645.1 | |||
| XM_017003900.1 | XP_016859389.1 | |||
| XM_017003901.1 | XP_016859390.1 | |||
| AF198489.1 | AAF32276.1 | |||
| AF411210.1 | AAM22616.1 | |||
| AK293855.1 | ||||
| AK296982.1 | ||||
| AK312950.1 | ||||
| BC067519.1 | AAH67519.1 | |||
| BC067520.1 | AAH67520.1 | |||
| BC067521.1 | AAH67521.1 | |||
| CD559574.2 |
More Information
Set Membership: |
|

Genomic Map