Assay Details
Target Gene Details
Entrez Gene ID: | 51599 |
Gene Name: | lipolysis stimulated lipoprotein receptor |
Gene Aliases: |
ILDR3, LISCH7 |
Location: |
Chr.19:35248656-35267964 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LSR | NM_001260489.1 | 1 | 92 | NP_001247418.1 |
| NM_001260490.1 | 1 | 92 | NP_001247419.1 | |
| NM_015925.6 | 1 | 92 | NP_057009.3 | |
| NM_205834.3 | 1 | 92 | NP_991403.1 | |
| NM_205835.3 | 1 | 92 | NP_991404.1 | |
| XM_005258980.1 | 1 | 67 | XP_005259037.1 | |
| XM_005258982.1 | 1 | 67 | XP_005259039.1 | |
| XM_011527026.1 | 1 | 67 | XP_011525328.1 | |
| AK126834.1 | 1 | 356 | BAC86714.1 | |
| BC047376.1 | AAH47376.2 | |||
| BM837160.1 | ||||
| BQ711340.1 | 1 | 92 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644228 | Chr.19:35209059 - 35249028 on Build GRCh38 | Gain |
|
| nsv1066553 | Chr.19:35207735 - 35258660 on Build GRCh38 | Gain |
|
| esv2758758 | Chr.19:35220175 - 35396211 on Build GRCh38 | Loss |
|
| nsv953283 | Chr.19:35247998 - 35314097 on Build GRCh38 | Deletion |
|
| nsv510765 | Chr.19:35217323 - 35399766 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map