Assay Details
Target Gene Details
Entrez Gene ID: | 29114 |
Gene Name: | transgelin 3 |
Gene Aliases: |
NP22, NP24, NP25 |
Location: |
Chr.3:111998739-112013888 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAGLN3 | NM_001008272.1 | NP_001008273.1 | ||
| NM_001008273.1 | NP_001008274.1 | |||
| NM_013259.2 | NP_037391.2 | |||
| AF087855.1 | AAP97165.1 | |||
| AF112201.1 | AAF17189.1 | |||
| AF303058.1 | AAL09330.1 | |||
| AK315289.1 | ||||
| BC015329.1 | AAH15329.1 | |||
| BG912462.1 | ||||
| BI752099.1 | ||||
| BI822856.1 | ||||
| BQ640029.1 | ||||
| BT007408.1 | AAP36076.1 | |||
| CK003985.1 |
More Information
Set Membership: |
|

Genomic Map