Assay Details
Target Gene Details
Entrez Gene ID: | 55075 |
Gene Name: | uveal autoantigen with coiled-coil domains and ankyrin repeats |
Gene Aliases: |
NUCLING |
Location: |
Chr.15:70654554-70778823 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UACA | NM_001008224.1 | 16 | 2700 | NP_001008225.1 |
| NM_018003.2 | 16 | 2477 | NP_060473.2 | |
| XM_005254529.4 | 15 | 2694 | XP_005254586.1 | |
| XM_011521752.2 | 16 | 2806 | XP_011520054.2 | |
| XM_011521753.2 | 15 | 2774 | XP_011520055.2 | |
| XM_011521754.2 | 15 | 2255 | XP_011520056.1 | |
| XM_017022394.1 | 15 | 2526 | XP_016877883.1 | |
| XM_017022395.1 | 16 | 2337 | XP_016877884.1 | |
| AB046781.2 | 16 | 2373 | BAB13387.2 | |
| AF322916.1 | 16 | 2422 | AAG49577.1 | |
| AK000990.1 | 1 | 438 | BAA91457.1 | |
| BC113407.1 | 16 | 2396 | ||
| BC113409.1 | 16 | 2396 | ||
| BC143266.1 | 16 | 2396 | ||
| BC143267.1 | 13 | 2069 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1594 | Chr.15:70645257 - 70677701 on Build GRCh38 | Insertion |
|
| nsv833049 | Chr.15:70582449 - 70687237 on Build GRCh38 | Loss |
|
| nsv1035437 | Chr.15:70644099 - 70746740 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114734080] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map