Assay Details
Target Gene Details
Entrez Gene ID: | 27030 |
Gene Name: | mutL homolog 3 |
Gene Aliases: |
HNPCC7 |
Location: |
Chr.14:75013764-75051532 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLH3 | NM_001040108.1 | NP_001035197.1 | ||
| NM_014381.2 | NP_055196.2 | |||
| XM_005267532.4 | XP_005267589.1 | |||
| XM_005267533.4 | XP_005267590.1 | |||
| XM_005267534.2 | XP_005267591.1 | |||
| XM_006720116.3 | XP_006720179.1 | |||
| XM_011536646.2 | XP_011534948.1 | |||
| XM_017021219.1 | XP_016876708.1 | |||
| AB039667.1 | BAA92353.1 | |||
| AF195657.1 | AAF23904.1 | |||
| AK298777.1 | ||||
| AL833875.1 | CAD38732.1 | |||
| BC036866.2 | AAH36866.1 | |||
| BC112167.1 | AAI12168.1 | |||
| L40399.1 | AAC42005.1 |
More Information
Set Membership: |
|

Genomic Map