Assay Details
Target Gene Details
Entrez Gene ID: | 2694 |
Gene Name: | gastric intrinsic factor |
Gene Aliases: |
IF, IFMH, INF, TCN3 |
Location: |
Chr.11:59829268-59845529 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GIF | NM_005142.2 | NP_005133.2 | ||
| XM_011544939.2 | XP_011543241.1 | |||
| AK301295.1 | ||||
| AK314275.1 | ||||
| BC037958.1 | AAH37958.1 | |||
| X76562.1 | CAA54061.1 |
More Information
Set Membership: |
|

Genomic Map