Assay Details
Target Gene Details
Entrez Gene ID: | 4007 |
Gene Name: | prickle planar cell polarity protein 3 |
Gene Aliases: |
LMO6 |
Location: |
Chr.X:49174802-49186400 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRICKLE3 | NM_001307979.1 | 8 | 1091 | NP_001294908.1 |
| NM_006150.4 | 8 | 1168 | NP_006141.2 | |
| AJ011654.1 | 8 | 1091 | CAA09726.1 | |
| AK299407.1 | ||||
| AK300866.1 | 7 | 993 | ||
| AK303200.1 | 8 | 1074 | ||
| AK303308.1 | 7 | 1052 | ||
| AK314788.1 | 8 | 1119 | ||
| BC002468.2 | 8 | 1121 | AAH02468.1 | |
| BC016856.1 | 8 | 1137 | AAH16856.1 | |
| BT007423.1 | 8 | 1065 | AAP36091.1 | |
| CR457211.1 | 8 | 1065 | CAG33492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map