Assay Details
Target Gene Details
Entrez Gene ID: | 51676 |
Gene Name: | ankyrin repeat and SOCS box containing 2 |
Gene Aliases: |
ASB-2 |
Location: |
Chr.14:93934153-93976796 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB2 | NM_001202429.1 | 8 | 1649 | NP_001189358.1 |
| NM_016150.4 | 6 | 1624 | NP_057234.2 | |
| XM_005267758.3 | 9 | 1709 | XP_005267815.1 | |
| XM_011536834.2 | 7 | 1559 | XP_011535136.1 | |
| XM_011536835.2 | 6 | 1198 | XP_011535137.1 | |
| XM_017021369.1 | 7 | 1608 | XP_016876858.1 | |
| AB056723.1 | 6 | 1624 | BAB64532.1 | |
| AB488462.1 | 8 | 1649 | ||
| AF159164.1 | 6 | 1009 | AAD45345.1 | |
| AJ251238.1 | 6 | 1278 | CAC17765.1 | |
| AK002049.1 | 6 | 1440 | ||
| AK057078.1 | 9 | 1597 | ||
| AK093271.1 | 1 | 1050 | ||
| AK303686.1 | 8 | 1468 | ||
| AK315628.1 | 6 | 1250 | ||
| AL137735.1 | 6 | 1045 | CAB70899.1 | |
| BC032354.1 | 6 | 1257 | AAH32354.1 |
More Information
Set Membership: |
|

Genomic Map