Assay Details
Target Gene Details
Entrez Gene ID: | 148808 |
Gene Name: | major facilitator superfamily domain containing 4A |
Gene Aliases: |
MFSD4, UNQ3064 |
Location: |
Chr.1:205568984-205602918 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD4A | NM_181644.4 | NP_857595.3 | ||
| XM_011509229.2 | XP_011507531.1 | |||
| AK090831.1 | ||||
| AK091896.1 | BAC03767.1 | |||
| AK293816.1 | ||||
| AK295148.1 | ||||
| AK304109.1 | ||||
| AK304120.1 | ||||
| AL713676.1 | CAD28481.1 | |||
| AY358107.1 | AAQ88474.1 | |||
| BC036549.2 | AAH36549.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549044 | Chr.1:205556670 - 205599085 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map