Assay Details
Target Gene Details
Entrez Gene ID: | 4853 |
Gene Name: | notch 2 |
Gene Aliases: |
AGS2, HJCYS, hN2 |
Location: |
Chr.1:119911553-120073449 on Build GRCh38 |
Assay Gene Location: | Within Exon 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOTCH2 | NM_024408.3 | 34 | 6681 | NP_077719.2 |
| XM_005270901.3 | 35 | 8995 | XP_005270958.1 | |
| XM_011541519.2 | 34 | 11789 | XP_011539821.1 | |
| XM_011541520.2 | 34 | 8699 | XP_011539822.1 | |
| XM_017001372.1 | 34 | 6351 | XP_016856861.1 | |
| XM_017001373.1 | 34 | 6330 | XP_016856862.1 | |
| AF308601.1 | 34 | 6396 | AAA36377.2 | |
| AF315356.1 | 34 | 6396 | AAG37073.1 | |
| AK027869.1 | 8 | 1520 | BAB55419.1 | |
| AL133036.1 | 1 | 358 | CAB61366.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2610 | Chr.1:119908437 - 119940512 on Build GRCh38 | Deletion |
|
| nsv831226 | Chr.1:119788976 - 119980845 on Build GRCh38 | Gain+Loss |
|
| nsv436325 | Chr.1:119848291 - 119991491 on Build GRCh38 | Deletion |
|
| nsv547625 | Chr.1:119880082 - 119923485 on Build GRCh38 | Loss |
|
| nsv524681 | Chr.1:118890895 - 121570001 on Build GRCh38 | Gain |
|
| nsv428201 | Chr.1:119724690 - 120154589 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map