Assay Details
Target Gene Details
Entrez Gene ID: | 261726 |
Gene Name: | TOR signaling pathway regulator |
Gene Aliases: |
TIP, TIP41, TIPRL1 |
Location: |
Chr.1:168178845-168202114 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIPRL | NM_001031800.2 | 1 | 247 | NP_001026970.1 |
| NM_152902.4 | 1 | 247 | NP_690866.1 | |
| AB097034.1 | 1 | 130 | BAC77387.1 | |
| AK313520.1 | 1 | 247 | ||
| AL049670.1 | 1 | 159 | CAB41244.1 | |
| BC009506.2 | 1 | 37 | AAH09506.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32660 | Chr.1:167825845 - 168220986 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map