Assay Details
Target Gene Details
Entrez Gene ID: | 64859 |
Gene Name: | nucleic acid binding protein 1 |
Gene Aliases: |
OBFC2A, SOSS-B2, SSB2 |
Location: |
Chr.2:191678072-191688522 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NABP1 | NM_001031716.2 | 1 | 521 | NP_001026886.1 |
| NM_001254736.1 | NP_001241665.1 | |||
| NR_045622.1 | ||||
| NR_045623.1 | 1 | 520 | ||
| XM_017004722.1 | 1 | 521 | XP_016860211.1 | |
| AK026486.1 | 1 | 520 | BAB15491.1 | |
| AL832659.1 | CAH56209.1 | |||
| AL833684.1 | ||||
| BC017114.1 | 1 | 124 | AAH17114.1 | |
| BC107723.1 | AAI07724.1 | |||
| BI553898.1 | ||||
| DA026631.1 | ||||
| DB477793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834494 | Chr.2:191673976 - 191903664 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map