Assay Details
Target Gene Details
Entrez Gene ID: | 8563 |
Gene Name: | THO complex 5 |
Gene Aliases: |
C22orf19, Fmip, PK1.3, fSAP79 |
Location: |
Chr.22:29508167-29554254 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THOC5 | NM_001002877.1 | 21 | 2497 | NP_001002877.1 |
| NM_001002878.1 | 21 | 2628 | NP_001002878.1 | |
| NM_001002879.1 | 20 | 2512 | NP_001002879.1 | |
| NM_003678.4 | 20 | 2381 | NP_003669.4 | |
| XM_005261797.1 | 21 | 2427 | XP_005261854.1 | |
| XM_005261798.1 | 22 | 2543 | XP_005261855.1 | |
| XM_005261799.1 | 21 | 2570 | XP_005261856.1 | |
| XM_005261801.3 | 20 | 2487 | XP_005261858.1 | |
| XM_006724347.1 | 22 | 2657 | XP_006724410.1 | |
| XM_017029027.1 | 22 | 3029 | XP_016884516.1 | |
| AB023200.1 | 20 | 2348 | ||
| BQ184247.1 | 1 | 67 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834172 | Chr.22:29503047 - 29661959 on Build GRCh38 | Loss |
|
| esv3647538 | Chr.22:29506399 - 29525757 on Build GRCh38 | Gain |
|
| esv3647536 | Chr.22:29477608 - 29543952 on Build GRCh38 | Gain |
|
| esv3893473 | Chr.22:29474962 - 29528836 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map