Assay Details
Target Gene Details
Entrez Gene ID: | 2151 |
Gene Name: | coagulation factor II thrombin receptor like 2 |
Gene Aliases: |
PAR-3, PAR3 |
Location: |
Chr.5:76615482-76623434 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F2RL2 | NM_001256566.1 | 2 | 1816 | NP_001243495.1 |
| NM_004101.3 | 2 | 1967 | NP_004092.1 | |
| BX537386.1 | 2 | 1948 | CAD97628.1 |
Target Gene Details
Entrez Gene ID: | 10788 |
Gene Name: | IQ motif containing GTPase activating protein 2 |
Gene Aliases: |
- |
Location: |
Chr.5:76403255-76708132 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQGAP2 | NM_001285460.1 | NP_001272389.1 | ||
| NM_001285461.1 | NP_001272390.1 | |||
| NM_001285462.1 | NP_001272391.1 | |||
| NM_006633.3 | NP_006624.2 | |||
| XM_005248410.2 | XP_005248467.1 | |||
| XM_005248414.1 | XP_005248471.1 | |||
| XM_011543108.1 | XP_011541410.1 | |||
| XM_017008960.1 | XP_016864449.1 | |||
| AB208856.1 | BAD92093.1 | |||
| AK291066.1 | ||||
| AK302510.1 | ||||
| AK302599.1 | ||||
| AK302981.1 | ||||
| AK303054.1 | ||||
| AK307555.1 | ||||
| AK311619.1 | ||||
| AY351902.1 | AAQ81291.1 | |||
| DB071610.1 | ||||
| DB075371.1 | ||||
| U51903.1 | AAB37765.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1022032 | Chr.5:76489281 - 76618178 on Build GRCh38 | Gain |
|
| dgv9884n54 | Chr.5:76528140 - 76618904 on Build GRCh38 | Loss |
|
| dgv5702n100 | Chr.5:76535212 - 76620080 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map