Assay Details
Target Gene Details
Entrez Gene ID: | 121391 |
Gene Name: | keratin 74 |
Gene Aliases: |
ADWH, HTSS2, HYPT3, K6IRS4, KRT5C, KRT6IRS4 |
Location: |
Chr.12:52565819-52573825 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT74 | NM_175053.3 | 9 | 2144 | NP_778223.2 |
| AF086480.1 | ||||
| AJ508777.1 | 9 | 2144 | CAD48514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv988599 | Chr.12:52563886 - 52568558 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114893604] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map