Assay Details
Target Gene Details
Entrez Gene ID: | 347468 |
Gene Name: | olfactory receptor family 13 subfamily H member 1 |
Gene Aliases: |
ORX1 |
Location: |
Chr.X:131544074-131545000 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR13H1 | NM_001004486.1 | 1 | 679 | NP_001004486.1 |
| BC137050.1 | 1 | 777 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528179 | Chr.X:131166643 - 131546956 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map