Assay Details
Target Gene Details
Entrez Gene ID: | 55735 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C11 |
Gene Aliases: |
dJ126A5.1 |
Location: |
Chr.1:6634168-6701906 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC11 | NM_018198.3 | 14 | 1542 | NP_060668.2 |
| XM_017001745.1 | 13 | 1395 | XP_016857234.1 | |
| AF306695.1 | 14 | 1456 | ||
| AK001599.1 | 14 | 1451 | BAA91780.1 | |
| AK025491.1 | 4 | 216 | ||
| AK093709.1 | 2 | 697 | ||
| AK095386.1 | 5 | 1264 | ||
| AK294539.1 | 13 | 1365 | ||
| AK298378.1 | 14 | 1428 | ||
| AL109978.1 | CAB53376.1 | |||
| AL833841.1 | 14 | 1416 | CAD38701.1 | |
| AY336749.1 | AAR02411.1 | |||
| BC006086.1 | 14 | 1438 | AAH06086.1 | |
| BC008772.1 | 14 | 1443 | AAH08772.1 | |
| BC014145.2 | 13 | 1294 | AAH14145.1 |
More Information
Set Membership: |
|

Genomic Map