| esv2422427 | 
						
							
								Chr.16:10001 - 2723164
							    on Build GRCh38
						
							
						 | 
						Duplication | 
						
							
								 NPRL3
							
								 IGFALS
							
								 MIR662
							
								 ZNF598
							
								 UBE2I
							
								 SNHG19
							
								 PRSS29P
							
								 TPSB2
							
								 TPSD1
							
								 PRSS27
							
								 MRPS34
							
								 FBXL16
							
								 NARFL
							
								 PRR35
							
								 CASKIN1
							
								 WDR90
							
								 NME4
							
								 GFER
							
								 MIR3180-5
							
								 MEIOB
							
								 NUBP2
							
								 WDR24
							
								 NPW
							
								 E4F1
							
								 MIR4717
							
								 SSTR5
							
								 POLR3K
							
								 LINC00254
							
								 TSR3
							
								 ERVK13-1
							
								 RNF151
							
								 C16orf59
							
								 UNKL
							
								 HAGHL
							
								 MIR4516
							
								 MAPK8IP3
							
								 MSLN
							
								 CHTF18
							
								 HBA2
							
								 LOC105371038
							
								 SNORA78
							
								 RGS11
							
								 LOC107984876
							
								 LOC105371184
							
								 RAB11FIP3
							
								 LOC105371045
							
								 METTL26
							
								 SYNGR3
							
								 HN1L
							
								 TBL3
							
								 RNPS1
							
								 WFIKKN1
							
								 HAGH
							
								 RHOT2
							
								 FAM173A
							
								 RAB40C
							
								 MIR3677
							
								 RPL3L
							
								 SNORD60
							
								 NME3
							
								 HBA1
							
								 PRR25
							
								 C16orf91
							
								 LOC101929280
							
								 CCNF
							
								 GNPTG
							
								 CEMP1
							
								 TMEM204
							
								 DDX11L10
							
								 RPL23AP5
							
								 ATP6V0C
							
								 PIGQ
							
								 MIR3177
							
								 RHBDL1
							
								 BRICD5
							
								 DECR2
							
								 AMDHD2
							
								 IFT140
							
								 NOXO1
							
								 ECI1
							
								 MIR6768
							
								 SNORA10
							
								 JMJD8
							
								 MIR1225
							
								 AXIN1
							
								 HBZ
							
								 LOC100134368
							
								 NTN3
							
								 FAM234A
							
								 HBM
							
								 LOC105371047
							
								 TSC2
							
								 MIR3176
							
								 DNASE1L2
							
								 BAIAP3
							
								 MIR5587
							
								 PDIA2
							
								 TMEM8A
							
								 SLC9A3R2
							
								 CACNA1H
							
								 MIR3178
							
								 CRAMP1
							
								 TPSG1
							
								 HS3ST6
							
								 RPS2
							
								 CCDC154
							
								 LOC652276
							
								 EME2
							
								 LINC00235
							
								 C1QTNF8
							
								 PGP
							
								 ARHGDIG
							
								 SNORA64
							
								 NTHL1
							
								 SSTR5-AS1
							
								 LUC7L
							
								 PDPK1
							
								 MIR940
							
								 CLCN7
							
								 RHBDF1
							
								 WASIR2
							
								 RAB26
							
								 LOC105371046
							
								 MRPL28
							
								 MCRIP2
							
								 FLJ42627
							
								 TRAF7
							
								 PKD1
							
								 MIR6511B1
							
								 TELO2
							
								 LMF1-AS1
							
								 NDUFB10
							
								 LOC106660606
							
								 MIR6859-4
							
								 SNHG9
							
								 ABCA17P
							
								 FAHD1
							
								 METRN
							
								 PTX4
							
								 CAPN15
							
								 LOC107987233
							
								 MIR6767
							
								 LOC729652
							
								 SPSB3
							
								 SNRNP25
							
								 NHLRC4
							
								 GNG13
							
								 MPG
							
								 SOX8
							
								 LMF1
							
								 LOC105371049
							
								 STUB1
							
								 TPSAB1
							
								 KCTD5
							
								 ABCA3
							
								 MSRB1
							
								 LOC100287175
							
								 TBC1D24
							
								 CCDC78
							
								 MLST8
							
								 HBQ1
							
								 RPUSD1
							
						 | 
					
				
		    		
						| nsv521569 | 
						
							
								Chr.16:1971892 - 2035996
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 NPW
							
								 SLC9A3R2
							
								 ZNF598
							
								 SYNGR3
							
								 GFER
							
								 TBL3
							
								 NOXO1
							
						 | 
					
				
		    		
						| dgv4858n54 | 
						
							
								Chr.16:1955562 - 2038584
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 NPW
							
								 SLC9A3R2
							
								 SNORA10
							
								 ZNF598
							
								 SYNGR3
							
								 RPS2
							
								 TBL3
							
								 RNF151
							
								 NDUFB10
							
								 SNORA64
							
								 SNHG9
							
								 GFER
							
								 NOXO1
							
								 SNORA78
							
						 | 
					
				
		    		
						| nsv952901 | 
						
							
								Chr.16:1888800 - 2135899
							    on Build GRCh38
						
							
						 | 
						Deletion | 
						
							
								 NPW
							
								 SLC9A3R2
							
								 SNORA10
							
								 MIR1225
							
								 ZNF598
							
								 HS3ST6
							
								 SYNGR3
							
								 RPS2
							
								 TBL3
							
								 RNF151
							
								 TSC2
							
								 PKD1
							
								 MIR6511B1
							
								 NDUFB10
							
								 LOC105371049
							
								 SNORA64
							
								 SNHG9
							
								 NTHL1
							
								 MSRB1
							
								 MIR4516
							
								 GFER
							
								 NOXO1
							
								 SNORA78
							
								 RPL3L
							
						 | 
					
				
		    		
						| nsv1160329 | 
						
							
								Chr.16:1961727 - 2128574
							    on Build GRCh38
						
							
						 | 
						Deletion | 
						
							
								 NPW
							
								 SLC9A3R2
							
								 SNORA10
							
								 MIR1225
							
								 ZNF598
							
								 SYNGR3
							
								 RPS2
							
								 TBL3
							
								 RNF151
							
								 TSC2
							
								 PKD1
							
								 MIR6511B1
							
								 NDUFB10
							
								 LOC105371049
							
								 SNORA64
							
								 SNHG9
							
								 NTHL1
							
								 GFER
							
								 NOXO1
							
								 SNORA78
							
						 | 
					
				
		    		
						| dgv4860n54 | 
						
							
								Chr.16:1974897 - 1980689
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 TBL3
							
								 NOXO1
							
						 | 
					
				
		    		
						| nsv471070 | 
						
							
								Chr.16:1947613 - 2024218
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 NPW
							
								 SNORA10
							
								 ZNF598
							
								 SYNGR3
							
								 RPS2
							
								 TBL3
							
								 RNF151
							
								 NDUFB10
							
								 SNORA64
							
								 SNHG9
							
								 GFER
							
								 NOXO1
							
								 SNORA78
							
								 RPL3L
							
						 | 
					
				
		    		
						| nsv833120 | 
						
							
								Chr.16:1893102 - 1980660
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 NDUFB10
							
								 SNORA10
							
								 SNORA64
							
								 SNHG9
							
								 MSRB1
							
								 HS3ST6
							
								 RPS2
							
								 TBL3
							
								 RNF151
							
								 NOXO1
							
								 SNORA78
							
								 RPL3L
							
						 | 
					
				
		    		
						| dgv336n27 | 
						
							
								Chr.16:1965120 - 2038584
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 NPW
							
								 SLC9A3R2
							
								 SNHG9
							
								 ZNF598
							
								 SYNGR3
							
								 GFER
							
								 TBL3
							
								 RNF151
							
								 NOXO1
							
								 SNORA78
							
						 |