Assay Details
Target Gene Details
Entrez Gene ID: | 9729 |
Gene Name: | KIAA0408 |
Gene Aliases: |
- |
Location: |
Chr.6:127438406-127459390 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0408 | NM_014702.4 | 5 | 1834 | NP_055517.3 |
| AB007868.1 | 3 | 2227 | BAA23704.2 | |
| AK091424.1 | 6 | 2190 | ||
| AK125983.1 | 6 | 2343 | BAC86375.1 | |
| AK127114.1 | ||||
| BC018396.1 | 5 | 1672 | AAH18396.2 |
Target Gene Details
Entrez Gene ID: | 387104 |
Gene Name: | SOGA family member 3 |
Gene Aliases: |
C6orf174, dJ403A15.3 |
Location: |
Chr.6:127438406-127519355 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOGA3 | NM_001012279.2 | 11 | 5528 | NP_001012279.1 |
More Information
Set Membership: |
|

Genomic Map