Assay Details
Target Gene Details
Entrez Gene ID: | 23272 |
Gene Name: | family with sequence similarity 208 member A |
Gene Aliases: |
C3orf63, RAP140, se89-1 |
Location: |
Chr.3:56620132-56683242 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM208A | NM_001112736.1 | 24 | 5949 | NP_001106207.1 |
| NM_015224.3 | 17 | 4900 | NP_056039.2 | |
| XM_005264999.1 | 24 | 5807 | XP_005265056.1 | |
| XM_006713077.2 | 24 | 5684 | XP_006713140.1 | |
| AB029028.1 | 7 | 2753 | BAA83057.1 | |
| AF180425.2 | 17 | 5184 | AAD55098.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460557 | Chr.3:56560707 - 56627292 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map