Assay Details
Target Gene Details
Entrez Gene ID: | 199745 |
Gene Name: | THAP domain containing 8 |
Gene Aliases: |
- |
Location: |
Chr.19:36034984-36054785 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THAP8 | NM_152658.2 | 1 | 183 | NP_689871.1 |
| XM_005258623.2 | 1 | 206 | XP_005258680.1 | |
| XM_011526604.2 | 1 | 206 | XP_011524906.1 | |
| AK057453.1 | BAB71493.1 | |||
| AK093048.1 | 1 | 184 | BAC04034.1 | |
| JQ410992.1 | 1 | 183 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833815 | Chr.19:36028901 - 36134034 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map