Assay Details
Target Gene Details
Entrez Gene ID: | 51004 |
Gene Name: | coenzyme Q6, monooxygenase |
Gene Aliases: |
CGI-10, CGI10, COQ10D6 |
Location: |
Chr.14:73949934-73963112 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COQ6 | NM_182476.2 | 11 | 1362 | NP_872282.1 |
| NM_182480.2 | 11 | 1365 | NP_872286.2 | |
| XM_006720156.1 | 11 | 1406 | XP_006720219.1 | |
| XM_011536807.1 | 10 | 1226 | XP_011535109.1 | |
| XM_011536808.1 | 10 | 1153 | XP_011535110.1 | |
| XM_011536809.2 | 10 | 1300 | XP_011535111.1 | |
| XM_011536810.2 | XP_011535112.1 | |||
| XM_017021351.1 | 10 | 1052 | XP_016876840.1 | |
| XM_017021352.1 | 9 | 1938 | XP_016876841.1 | |
| AF132944.1 | 11 | 1288 | AAD27719.1 | |
| AK130167.1 | 11 | 1319 | ||
| AK222965.1 | 11 | 1304 | BAD96685.1 | |
| AK296040.1 | 11 | 1373 | ||
| BC014181.2 | 11 | 1311 | AAH14181.1 | |
| BC014483.1 | 11 | 1299 | AAH14483.1 | |
| BX248000.1 | 10 | 1300 | CAD62332.1 | |
| BX648434.1 | 4 | 2880 |
Target Gene Details
Entrez Gene ID: | 957 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 5 |
Gene Aliases: |
CD39L4, NTPDase-5, PCPH |
Location: |
Chr.14:73959420-74019326 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD5 | NM_001321984.1 | NP_001308913.1 | ||
| XM_006720325.3 | XP_006720388.1 | |||
| XM_017021814.1 | XP_016877303.1 | |||
| XM_017021815.1 | XP_016877304.1 | |||
| XM_017021817.1 | XP_016877306.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1352 | Chr.14:73951320 - 73996025 on Build GRCh38 | Deletion |
|
| nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain |
|
| dgv1937n100 | Chr.14:73950133 - 74115803 on Build GRCh38 | Gain |
|
| nsv524396 | Chr.14:73958235 - 74023032 on Build GRCh38 | Gain |
|
| nsv1046028 | Chr.14:73957079 - 74032042 on Build GRCh38 | Loss |
|
| nsv1051583 | Chr.14:73953737 - 73995987 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map