Assay Details
Target Gene Details
Entrez Gene ID: | 148898 |
Gene Name: | ZNF436 antisense RNA 1 |
Gene Aliases: |
C1orf213 |
Location: |
Chr.1:23368971-23371839 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF436-AS1 | NR_033690.1 | 1 | 2291 | |
| NR_033691.1 | ||||
| AK074989.1 | 1 | 2205 | BAC11338.1 | |
| BC028411.1 | 1 | 2032 | AAH28411.1 | |
| BC054115.1 | 1 | 2295 | AAH54115.1 | |
| BC068594.1 | 1 | 2273 | AAH68594.1 | |
| BX098506.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525446 | Chr.1:23369659 - 23374819 on Build GRCh38 | Gain |
|

Genomic Map