Assay Details
Target Gene Details
Entrez Gene ID: | 1017 |
Gene Name: | cyclin dependent kinase 2 |
Gene Aliases: |
CDKN2, p33(CDK2) |
Location: |
Chr.12:55966769-55972789 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK2 | NM_001290230.1 | NP_001277159.1 | ||
| NM_001798.4 | NP_001789.2 | |||
| NM_052827.3 | NP_439892.2 | |||
| XM_011537732.1 | XP_011536034.1 | |||
| AB012305.1 | BAA32794.1 | |||
| AK291941.1 | ||||
| AK293246.1 | ||||
| BC003065.2 | AAH03065.1 | |||
| BJ991087.1 | ||||
| BT006821.1 | AAP35467.1 | |||
| DA814453.1 | ||||
| M68520.1 | AAA35667.1 | |||
| X61622.1 | CAA43807.1 | |||
| X62071.1 | CAA43985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832424 | Chr.12:55881442 - 56073133 on Build GRCh38 | Loss |
|
| nsv1051961 | Chr.12:55868436 - 56235643 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map