Assay Details
Target Gene Details
Entrez Gene ID: | 10279 |
Gene Name: | protease, serine 16 |
Gene Aliases: |
TSSP |
Location: |
Chr.6:27247701-27256620 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS16 | NM_005865.3 | NP_005856.1 | ||
| XM_017010161.1 | XP_016865650.1 | |||
| XM_017010162.1 | XP_016865651.1 | |||
| XM_017010163.1 | XP_016865652.1 | |||
| XM_017010164.1 | XP_016865653.1 | |||
| XM_017010165.1 | XP_016865654.1 | |||
| AF052514.1 | AAC33563.1 | |||
| AK093590.1 | ||||
| AK097918.1 | ||||
| AK098033.1 | ||||
| AK126160.1 | ||||
| AK226091.1 | ||||
| AK303106.1 | ||||
| AK314037.1 |
More Information
Set Membership: |
|

Genomic Map