Assay Details
Target Gene Details
Entrez Gene ID: | 2695 |
Gene Name: | gastric inhibitory polypeptide |
Gene Aliases: |
- |
Location: |
Chr.17:48958556-48968593 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GIP | NM_004123.2 | NP_004114.1 | ||
| BC069100.1 | AAH69100.1 | |||
| BC069663.1 | AAH69663.1 | |||
| BC069686.1 | AAH69686.1 | |||
| BC069746.1 | AAH69746.1 | |||
| BC096146.1 | AAH96146.1 | |||
| BC096147.1 | AAH96147.1 | |||
| BC096148.3 | AAH96148.1 | |||
| BC096149.3 | AAH96149.1 | |||
| E02006.1 | ||||
| M18185.1 | AAA88043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2073 | Chr.17:48928271 - 48962703 on Build GRCh38 | Insertion |
|
| nsv482204 | Chr.17:48958556 - 48968593 on Build GRCh38 | Loss |
|
| esv2651898 | Chr.17:48959344 - 48961248 on Build GRCh38 | Deletion |
|
| nsv833472 | Chr.17:48895840 - 49078381 on Build GRCh38 | Gain |
|
| nsv482969 | Chr.17:48843507 - 48994527 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map