Assay Details
Target Gene Details
Entrez Gene ID: | 5970 |
Gene Name: | RELA proto-oncogene, NF-kB subunit |
Gene Aliases: |
NFKB3, p65 |
Location: |
Chr.11:65653596-65662972 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RELA | NM_001145138.1 | NP_001138610.1 | ||
| NM_001243984.1 | NP_001230913.1 | |||
| NM_001243985.1 | NP_001230914.1 | |||
| NM_021975.3 | NP_068810.3 | |||
| XM_011545206.1 | XP_011543508.1 | |||
| XM_011545207.1 | XP_011543509.1 | |||
| AK303262.1 | ||||
| AK308020.1 | ||||
| AK308478.1 | ||||
| BC011603.1 | AAH11603.1 | |||
| BC014095.2 | AAH14095.1 | |||
| BC033522.1 | AAH33522.1 | |||
| BC110830.1 | AAI10831.1 | |||
| L19067.1 | AAA20946.1 | |||
| M62399.1 | AAA36408.1 | |||
| U33838.1 | AAA92888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1969n54 | Chr.11:65568234 - 65663889 on Build GRCh38 | Loss |
|
| nsv951018 | Chr.11:65525430 - 65662029 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map