Assay Details
Target Gene Details
Entrez Gene ID: | 23650 |
Gene Name: | tripartite motif containing 29 |
Gene Aliases: |
ATDC |
Location: |
Chr.11:120111286-120138179 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM29 | NM_012101.3 | 9 | 2872 | NP_036233.2 |
| XM_005271490.1 | 9 | 2049 | XP_005271547.1 | |
| XM_011542729.1 | 10 | 2998 | XP_011541031.1 | |
| XM_011542732.1 | 10 | 2995 | XP_011541034.1 | |
| XM_011542733.1 | 10 | 2151 | XP_011541035.1 | |
| XM_017017453.1 | 10 | 2307 | XP_016872942.1 | |
| XM_017017454.1 | 9 | 2205 | XP_016872943.1 | |
| AF230388.1 | 9 | 2872 | AAG50167.1 | |
| AF230389.1 | AAG50168.1 | |||
| AK093404.1 | 5 | 1942 | ||
| AK122720.1 | 10 | 2688 | ||
| AL832678.1 | 2 | 2814 | ||
| BC010644.1 | 9 | 2818 | ||
| BC017352.2 | 9 | 2811 | AAH17352.1 | |
| BX648072.1 | 1 | 3593 | ||
| L24203.1 | 9 | 2872 | AAA35762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050132 | Chr.11:120105602 - 120139729 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map