Assay Details
Target Gene Details
Entrez Gene ID: | 26659 |
Gene Name: | olfactory receptor family 7 subfamily A member 5 |
Gene Aliases: |
HTPCR2 |
Location: |
Chr.19:14825743-14835197 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR7A5 | NM_017506.1 | NP_059976.1 | ||
| XM_011527912.2 | 2 | 2152 | XP_011526214.1 | |
| XM_011527913.2 | 2 | 2078 | XP_011526215.1 | |
| XM_017026583.1 | 2 | 2041 | XP_016882072.1 |
Target Gene Details
Entrez Gene ID: | 26664 |
Gene Name: | olfactory receptor family 7 subfamily C member 1 |
Gene Aliases: |
CIT-HSP-146E8, HSTPCR86P, OR19-5, OR7C4, TPCR86 |
Location: |
Chr.19:14797550-14835376 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR7C1 | XM_005259853.3 | XP_005259910.1 | ||
| XM_011527915.2 | XP_011526217.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map