Assay Details
Target Gene Details
Entrez Gene ID: | 23205 |
Gene Name: | acyl-CoA synthetase bubblegum family member 1 |
Gene Aliases: |
BG, BG1, BGM, GR-LACS, LPD |
Location: |
Chr.15:78167468-78234707 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSBG1 | NM_001199377.1 | 10 | 1521 | NP_001186306.1 |
| NM_015162.4 | 10 | 1533 | NP_055977.3 | |
| XM_011521390.2 | 10 | 1347 | XP_011519692.1 | |
| XM_011521391.2 | 9 | 3206 | XP_011519693.2 | |
| XM_017022025.1 | 10 | 1367 | XP_016877514.1 | |
| XM_017022026.1 | 13 | 2168 | XP_016877515.1 | |
| AF179481.1 | 10 | 1374 | AAG09404.1 | |
| AK054882.1 | 10 | 1383 | ||
| AK294202.1 | 6 | 819 | ||
| AK295269.1 | 10 | 1339 | ||
| AK299597.1 | 9 | 1282 | ||
| AK314508.1 | 10 | 1533 | ||
| BC009289.2 | 10 | 1332 | AAH09289.1 |
More Information
Set Membership: |
|

Genomic Map