Assay Details
Target Gene Details
Entrez Gene ID: | 7105 |
Gene Name: | tetraspanin 6 |
Gene Aliases: |
T245, TM4SF6, TSPAN-6 |
Location: |
Chr.X:100627108-100637104 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN6 | NM_001278740.1 | NP_001265669.1 | ||
| NM_001278741.1 | NP_001265670.1 | |||
| NM_001278742.1 | NP_001265671.1 | |||
| NM_001278743.1 | NP_001265672.1 | |||
| NM_003270.3 | NP_003261.1 | |||
| XM_011531018.1 | XP_011529320.1 | |||
| AB097016.1 | BAC77369.1 | |||
| AB209876.1 | BAD93113.1 | |||
| AF043906.1 | AAC64257.1 | |||
| AF053453.1 | AAC69710.1 | |||
| AF133426.1 | AAF08365.1 | |||
| AK075072.1 | ||||
| AK299176.1 | ||||
| AK300852.1 | ||||
| AK309581.1 | ||||
| AK315773.1 | ||||
| AY358825.1 | AAQ89184.1 | |||
| BC012389.1 | AAH12389.1 | |||
| BT006977.1 | AAP35623.1 | |||
| CN274931.1 | ||||
| CR457115.1 | CAG33396.1 | |||
| CX868833.1 | ||||
| U84895.1 | AAD00560.1 |
More Information
Set Membership: |
|

Genomic Map