Assay Details
Target Gene Details
Entrez Gene ID: | 119548 |
Gene Name: | pancreatic lipase related protein 3 |
Gene Aliases: |
- |
Location: |
Chr.10:116427879-116477961 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNLIPRP3 | NM_001011709.2 | NP_001011709.2 | ||
| XM_011539276.1 | XP_011537578.1 | |||
| XM_011539277.1 | XP_011537579.1 | |||
| XM_011539278.1 | XP_011537580.1 | |||
| XM_011539279.1 | XP_011537581.1 | |||
| AL833418.1 | ||||
| BC117224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832001 | Chr.10:116378858 - 116555547 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map