Assay Details
Target Gene Details
Entrez Gene ID: | 340596 |
Gene Name: | lipoma HMGIC fusion partner-like 1 |
Gene Aliases: |
- |
Location: |
Chr.X:112630651-112680178 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL1 | NM_178175.3 | 4 | 1450 | NP_835469.1 |
| XM_011530943.2 | 4 | 1660 | XP_011529245.1 | |
| XM_011530944.2 | XP_011529246.1 | |||
| XM_011530946.2 | XP_011529248.1 | |||
| XM_017029485.1 | 4 | 1855 | XP_016884974.1 | |
| AK289946.1 | ||||
| AY217350.1 | 4 | 1357 | AAO60107.1 | |
| AY358208.1 | 3 | 1259 | AAQ88575.1 |

Genomic Map