Assay Details
Target Gene Details
Entrez Gene ID: | 145483 |
Gene Name: | family with sequence similarity 161 member B |
Gene Aliases: |
C14orf44, c14_5547 |
Location: |
Chr.14:73932031-73950414 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM161B | XM_011536475.1 | XP_011534777.1 | ||
| CR749537.1 |
Target Gene Details
Entrez Gene ID: | 57862 |
Gene Name: | zinc finger protein 410 |
Gene Aliases: |
APA-1, APA1 |
Location: |
Chr.14:73886615-73932288 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF410 | NM_001242924.1 | 14 | 2521 | NP_001229853.1 |
| NM_001242926.1 | 11 | 2211 | NP_001229855.1 | |
| NM_001242927.1 | 11 | 2133 | NP_001229856.1 | |
| NM_001242928.1 | 13 | 2445 | NP_001229857.1 | |
| NM_021188.2 | 12 | 2352 | NP_067011.1 | |
| NR_040251.1 | 12 | 2320 | ||
| AA652386.1 | ||||
| AK298307.1 | ||||
| BC050683.1 | 11 | 2021 | AAH50683.1 | |
| U90919.1 | 12 | 2084 | AAB51059.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain |
|
| dgv663e212 | Chr.14:73931191 - 73936946 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map