Assay Details
Target Gene Details
Entrez Gene ID: | 84313 |
Gene Name: | vacuolar protein sorting 25 homolog |
Gene Aliases: |
DERP9, EAP20, FAP20 |
Location: |
Chr.17:42773436-42779600 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS25 | NM_032353.3 | NP_115729.1 | ||
| XM_017025224.1 | XP_016880713.1 | |||
| AB014763.1 | BAB87804.1 | |||
| AK312092.1 | ||||
| BC006282.2 | AAH06282.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833450 | Chr.17:42660985 - 42825144 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map