Assay Details
Target Gene Details
Entrez Gene ID: | 55705 |
Gene Name: | importin 9 |
Gene Aliases: |
Imp9 |
Location: |
Chr.1:201829160-201884294 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPO9 | NM_018085.4 | NP_060555.2 | ||
| AF410465.1 | AAL01416.1 | |||
| AK027532.1 | BAB55181.1 | |||
| AK074740.1 | BAC11173.1 | |||
| BC003604.1 | AAH03604.2 |
Target Gene Details
Entrez Gene ID: | 102466724 |
Gene Name: | microRNA 6739 |
Gene Aliases: |
hsa-mir-6739 |
Location: |
Chr.1:201863373-201863447 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6739 | NR_106797.1 |
More Information
Set Membership: |
|

Genomic Map