Assay Details
Target Gene Details
Entrez Gene ID: | 126626 |
Gene Name: | GA binding protein transcription factor beta subunit 2 |
Gene Aliases: |
GABPB-2 |
Location: |
Chr.1:151070180-151125545 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map