Assay Details
Target Gene Details
Entrez Gene ID: | 389668 |
Gene Name: | XK related 9 |
Gene Aliases: |
XRG9 |
Location: |
Chr.8:70669365-71066635 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XKR9 | NM_001011720.1 | 1 | 41 | NP_001011720.1 |
| NM_001287258.1 | 1 | 41 | NP_001274187.1 | |
| NM_001287259.1 | 1 | 41 | NP_001274188.1 | |
| XM_006716447.3 | XP_006716510.1 | |||
| XM_011517524.2 | XP_011515826.1 | |||
| XM_011517526.2 | XP_011515828.1 | |||
| XM_011517527.2 | XP_011515829.1 | |||
| XM_017013405.1 | XP_016868894.1 | |||
| AY534247.1 | 1 | 41 | AAT07096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576585 | Chr.8:70645369 - 70750875 on Build GRCh38 | Gain |
|

Genomic Map