Assay Details
Target Gene Details
Entrez Gene ID: | 3998 |
Gene Name: | lectin, mannose binding 1 |
Gene Aliases: |
ERGIC-53, ERGIC53, F5F8D, FMFD1, MCFD1, MR60, gp58 |
Location: |
Chr.18:59327823-59359276 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMAN1 | NM_005570.3 | 13 | 2503 | NP_005561.1 |
| X71661.1 | 13 | 2492 | CAA50653.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv577051 | Chr.18:59313335 - 59354317 on Build GRCh38 | Loss |
|
| nsv522853 | Chr.18:59328164 - 59365421 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map